Learn More
Ace Glass, Inc. O-RING FETFE SIZE 235 O-RING FETFE SIZE 235
SDP
Catalog No. 50979554
Encompass_Preferred
Size 235 FETFE o-ring, Pack/3. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-979-554 Supplier Ace Glass, Inc. Supplier No. 7855764
Size 235 FETFE o-ring, Pack/3. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.