Learn More
Waters Corp O-Ring, FKM, BS019, 5 per pack
Catalog No. 502392301 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement O-ring is designed for REIMS systems.
  • Material: FKM (Viton)
  • Size: BS019
  • Quantity: 5 per pack
Catalog No. 50-239-2301 Supplier Waters Corp Supplier No. 700000455
This replacement O-ring is designed for REIMS systems.
  • Material: FKM (Viton)
  • Size: BS019
  • Quantity: 5 per pack