Learn More
Andwin Scientific O-RING FOR HIRSCHMANN PIPETUS
SDP
Catalog No. NC0831985
Encompass_Preferred
The Hirschmann Silicone Adapter is an accessory for the Pipetus Pipette Filler, which is a product that provides highly accurate pipetting. It can also be used for measuring, volumetric and Pasteur pipettes, from 0.1 mL to 200 mL. The adapter is sold separately from the pipetus pipette filler.
Catalog No. NC0831985 Supplier Andwin Scientific Supplier No. 9903205
The Hirschmann Silicone Adapter is an accessory for the Pipetus Pipette Filler, which is a product that provides highly accurate pipetting. It can also be used for measuring, volumetric and Pasteur pipettes, from 0.1 mL to 200 mL. The adapter is sold separately from the pipetus pipette filler.