Learn More
Glass Expansion O-ring for Sampler Cone.- Suitable for: Agilent 4500, 7500
SDP
Catalog No. NC1611703
Encompass_Preferred
O-ring for Sampler Cone (G1820-65025). Suitable for Agilent 4500 and 7500 ICP-MS sampler cones.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1611703 Supplier Glass Expansion Supplier No. AT5001
O-ring for Sampler Cone (G1820-65025). Suitable for Agilent 4500 and 7500 ICP-MS sampler cones.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001