Learn More
Markes International Ltd O-RING FOR MICRO-CHAMBER 4/PK
SDP
Catalog No. NC2090695
Encompass
For sample pan seal of high temperature Micro-Chamber (temperatures < 200 degree C)- pk 4
This item is not returnable. View return policy
Catalog No. NC2090695 Supplier Markes International Ltd Supplier No. MMCHOR250
For sample pan seal of high temperature Micro-Chamber (temperatures < 200 degree C)- pk 4