Learn More
Glass Expansion O-ring for PerkinElmer NexION 2200/5000 ICP-MS Hyper Skimmer Cone, PKT 5 (09211389) Replace if brittle, craked/damaged
SDP
Catalog No. 502702949
Encompass_Preferred
O-ring for PerkinElmer NexION 5000 ICP-MS Hyper Skimmer Cone, PKT 5 (09211389). Replace if brittle, cracked/damaged.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2949 Supplier Glass Expansion Supplier No. PE5125
O-ring for PerkinElmer NexION 5000 ICP-MS Hyper Skimmer Cone, PKT 5 (09211389). Replace if brittle, cracked/damaged.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001