Learn More
Ace Glass, Inc. O-Rings, Kalrez 100mm Flange
SDP
Catalog No. 50975678
Encompass
Kalrez o-ring, fits 100mm Duran Flange. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-975-678 Supplier Ace Glass, Inc. Supplier No. 7855696
Kalrez o-ring, fits 100mm Duran Flange. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.