Learn More
Ace Glass, Inc. O-Rings, Kalrez 200mm Flange
SDP
Catalog No. 50975680
Encompass
Kalrez o-ring, fits 200mm Duran Flange. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-975-680 Supplier Ace Glass, Inc. Supplier No. 7855698
Kalrez o-ring, fits 200mm Duran Flange. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.