Learn More
Chemglass Life Sciences O-Ring, Kalrez 4079, Size #114 - Sealing element for laboratory fittings and valves
SDP
Catalog No. NC2953685
Encompass_Preferred
Kalrez 4079 perfluoroelastomer O-ring, size #114, designed for high-temperature and chemically aggressive laboratory sealing applications. It combines elastomeric flexibility with PTFE-like chemical resistance for reliable long-term performance.
  • Provides long-term service to 316°C and intermittent service to 343°C.
  • Not recommended for sealing applications below -5°C.
  • Resistant to polar solvents, strong organic solvents, and common acids.
  • Compatible with oxidizers, metal halogen compounds, chlorine, and inorganic salt solutions.
  • Suitable for use with fuels and hydrocarbon-based fluids.
Catalog No. NC2953685 Supplier Chemglass Life Sciences Supplier No. CG308K114
Kalrez 4079 perfluoroelastomer O-ring, size #114, designed for high-temperature and chemically aggressive laboratory sealing applications. It combines elastomeric flexibility with PTFE-like chemical resistance for reliable long-term performance.
  • Provides long-term service to 316°C and intermittent service to 343°C.
  • Not recommended for sealing applications below -5°C.
  • Resistant to polar solvents, strong organic solvents, and common acids.
  • Compatible with oxidizers, metal halogen compounds, chlorine, and inorganic salt solutions.
  • Suitable for use with fuels and hydrocarbon-based fluids.