Learn More
Chemglass Life Sciences O-Ring, Kalrez 4079, #128
SDP
Catalog No. 501531586
Encompass_Preferred
O-Ring, Kalrez 4079, #128. Kalrez 4079 perfluroelastomer combines the elastomeric properties of Viton with the chemical resistance of PTFE. Kalrez 4079 exhibits exceptional thermal stability and is serviceable at long-term temperatures to 316C and intermittent temperatures to 343C. Kalrez 4079 is not recommended for use in sealing applications below -5C.
Catalog No. 50-153-1586 Supplier Chemglass Life Sciences Supplier No. CG308K128
O-Ring, Kalrez 4079, #128. Kalrez 4079 perfluroelastomer combines the elastomeric properties of Viton with the chemical resistance of PTFE. Kalrez 4079 exhibits exceptional thermal stability and is serviceable at long-term temperatures to 316C and intermittent temperatures to 343C. Kalrez 4079 is not recommended for use in sealing applications below -5C.