Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 007 O-RING KALREZ SIZE 007
SDP
Catalog No. 50979537
Encompass
Size 007 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-979-537 Supplier Ace Glass, Inc. Supplier No. 7855602
Size 007 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.