Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 105 O-RING KALREZ SIZE 105
SDP
Catalog No. 50975663
Encompass
Size 105 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-975-663 Supplier Ace Glass, Inc. Supplier No. 7855650
Size 105 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.