Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 112 O-RING KALREZ SIZE 112
SDP
Catalog No. 50979540
Encompass
Size 112 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-979-540 Supplier Ace Glass, Inc. Supplier No. 7855620
Size 112 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.