Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 122 O-RING KALREZ SIZE 122
SDP
Catalog No. 50979543
Encompass
Size 122 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-979-543 Supplier Ace Glass, Inc. Supplier No. 7855657
Size 122 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.