Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 128 O-RING KALREZ SIZE 128
SDP
Catalog No. 50979544
Encompass
Size 128 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-979-544 Supplier Ace Glass, Inc. Supplier No. 7855671
Size 128 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.