Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 211 O-RING KALREZ SIZE 211
SDP
Catalog No. 50979541
Encompass
Size 211 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-979-541 Supplier Ace Glass, Inc. Supplier No. 7855632
Size 211 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.