Learn More
Ace Glass, Inc. O-RING KALREZ SIZE 225 O-RING KALREZ SIZE 225
SDP
Catalog No. 50975662
Encompass
Size 225 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-975-662 Supplier Ace Glass, Inc. Supplier No. 7855644
Size 225 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.