Learn More
Bellco Glass, Inc. O-Ring Kit 7L
SDP
Catalog No. 501944962
Encompass_Preferred
O-ring replacement kit. MADE IN THE USA. Size: 7l A523-234 is for 3/8" accessories A523-407 is for 1/4" accessories A542-522 is for 5/16"Thermowell A542-521 is for 3/4" accessories A542-523 is the inner o-ring for the probe holder 1964-88803 is A542-528.
Catalog No. 50-194-4962 Supplier Bellco Glass, Inc. Supplier No. 196496070
O-ring replacement kit. MADE IN THE USA. Size: 7l A523-234 is for 3/8" accessories A523-407 is for 1/4" accessories A542-522 is for 5/16"Thermowell A542-521 is for 3/4" accessories A542-523 is the inner o-ring for the probe holder 1964-88803 is A542-528.