Learn More
Ace Glass, Inc. O-ring kit, FETFE, (12)each of sizes 008, 009, 010, 011, 012, 013, 014, 018, 108, 110, 111, 112, 116, 121, 136, 210, 212, 220
SDP
Catalog No. 50975709
Encompass
FETFE o-ring kit. FETFE has a good compression set, along with excellent temperature and chemical compatibility. Includes 12 each of sizes: 008, 009, 010, 011, 012, 013, 014, 018, 108, 110, 111, 112, 116, 121, 136, 210, 212, and 220.
Catalog No. 50-975-709 Supplier Ace Glass, Inc. Supplier No. 7855799
FETFE o-ring kit. FETFE has a good compression set, along with excellent temperature and chemical compatibility. Includes 12 each of sizes: 008, 009, 010, 011, 012, 013, 014, 018, 108, 110, 111, 112, 116, 121, 136, 210, 212, and 220.