Learn More
Glass Expansion O-ring Kit for Agilent D-Torch ICP-OES
SDP
Catalog No. 502703052
Encompass_Preferred
Replacement O-ring kit for Agilent D-Torch ICP-OES. Compatible with Glass Expansion D-Torches for Agilent 5000 SVDV/VDV/RV Series ICP-OES. D-Torch O-rings are recommended to be replaced at least once per year, or every 6 months if the D-Torch is frequently removed for cleaning and application changeovers. To help facilitate O-ring removal and installation, use the O-ring removal tool P/N 50-100-1636.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3052 Supplier Glass Expansion Supplier No. 708031500
Replacement O-ring kit for Agilent D-Torch ICP-OES. Compatible with Glass Expansion D-Torches for Agilent 5000 SVDV/VDV/RV Series ICP-OES. D-Torch O-rings are recommended to be replaced at least once per year, or every 6 months if the D-Torch is frequently removed for cleaning and application changeovers. To help facilitate O-ring removal and installation, use the O-ring removal tool P/N 50-100-1636.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001