Learn More
Glass Expansion O-ring kit for D-Torch Optima 2/4/5/7000DV & Optima 8X00
SDP
Catalog No. 502703086
Encompass_Preferred
O-Ring Kit for D-Torch Optima 2/4/5/7000DV & Optima 8x00 (N0777715). Four replacement O-rings for use with the following Glass Expansion D-Torches: Optima 2/4/5/7000DV 501907022, 30-808-2926; and Optima 8x00 501907027, 30-808-3300. Use O-ring removal tool 50-100-1636.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3086 Supplier Glass Expansion Supplier No. 708030762
O-Ring Kit for D-Torch Optima 2/4/5/7000DV & Optima 8x00 (N0777715). Four replacement O-rings for use with the following Glass Expansion D-Torches: Optima 2/4/5/7000DV 501907022, 30-808-2926; and Optima 8x00 501907027, 30-808-3300. Use O-ring removal tool 50-100-1636.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001