Learn More
Glass Expansion Sampler Cone O-ring, Pack of 5 (N8120511) Replace if brittle, craked/damaged
SDP
Catalog No. NC1585741
Encompass_Preferred
Sampler Cone O-Ring, PKT 5 (N8120511). Compatible with PerkinElmer Elan 6000/9000/DRCII/DRC-E ICP-MS. Replace if brittle, cracked/damaged.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1585741 Supplier Glass Expansion Supplier No. PE5011
Sampler Cone O-Ring, PKT 5 (N8120511). Compatible with PerkinElmer Elan 6000/9000/DRCII/DRC-E ICP-MS. Replace if brittle, cracked/damaged.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001