Learn More
Glass Expansion O-ring Kit Shimadzu 2030 ICP-MS Interface
SDP
Catalog No. 502703027
Encompass_Preferred
O-ring Kit Shimadzu 2030 ICP-MS Interface. Replaces S036-12553-01, S036-12553-05, S036-11348-21 and S036-11345-22. Use O-ring removal tool 50-100-1636.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3027 Supplier Glass Expansion Supplier No. 708031908
O-ring Kit Shimadzu 2030 ICP-MS Interface. Replaces S036-12553-01, S036-12553-05, S036-11348-21 and S036-11345-22. Use O-ring removal tool 50-100-1636.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001