Learn More
Chemglass Life Sciences 0-12mm Replacement O-Ring Kit, TFE Stem, Viton, To Consist Of: 1ea: #110 2ea: #112
SDP
Catalog No. CG53703
Encompass_Preferred
0-12mm Replacement O-Ring Kit, TFE Stem, Viton, To Consist Of: 1ea: #110 2ea: #112. Each kit contains a set of three o-rings, one for the tip and two for the barrel seal. O-ring sizes are listed in the following chart. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez.O-Ring Specifications:0-12mm: #110 Tip O-Ring, #112 Shaft O-Ring
Catalog No. CG53703 Supplier Chemglass Life Sciences Supplier No. CG53703
0-12mm Replacement O-Ring Kit, TFE Stem, Viton, To Consist Of: 1ea: #110 2ea: #112. Each kit contains a set of three o-rings, one for the tip and two for the barrel seal. O-ring sizes are listed in the following chart. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez.O-Ring Specifications:0-12mm: #110 Tip O-Ring, #112 Shaft O-Ring