Learn More
Chemglass Life Sciences 0-4mm Replacement O-Ring Kit, Glass Stem, Viton, To Consist Of: 1ea: #006 2ea: #009
SDP
Catalog No. CG58501
Encompass_Preferred
0-4mm Replacement O-Ring Kit, Glass Stem, Viton, To Consist Of: 1ea: #006 2ea: #009. Each kit contains a set of three o-rings, one for the tip and two for the barrel seal. O-ring sizes are listed in the following chart. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez.O-Ring Specifications:#006 Tip O-Ring#009 Shaft O-Ring
Catalog No. CG58501 Supplier Chemglass Life Sciences Supplier No. CG58501
0-4mm Replacement O-Ring Kit, Glass Stem, Viton, To Consist Of: 1ea: #006 2ea: #009. Each kit contains a set of three o-rings, one for the tip and two for the barrel seal. O-ring sizes are listed in the following chart. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez.O-Ring Specifications:#006 Tip O-Ring#009 Shaft O-Ring