Learn More
Chemglass Life Sciences 0-8mm Replacement O-Ring Kit, TFE Stem, Kalrez, To Consist Of: 1ea: #108 2ea: #110
SDP
Catalog No. CG53708
Encompass_Preferred
0-8mm Replacement O-Ring Kit, TFE Stem, Kalrez, To Consist Of: 1ea: #108 2ea: #110. Each kit contains a set of three o-rings, one for the tip and two for the barrel seal. O-ring sizes are listed in the following chart. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez.O-Ring Specifications:0-8mm: #108 Tip O-Ring, #110 Shaft O-Ring
Catalog No. CG53708 Supplier Chemglass Life Sciences Supplier No. CG53708
0-8mm Replacement O-Ring Kit, TFE Stem, Kalrez, To Consist Of: 1ea: #108 2ea: #110. Each kit contains a set of three o-rings, one for the tip and two for the barrel seal. O-ring sizes are listed in the following chart. If individual o-ring is required see CG-305 for Viton or CG-308 for Kalrez.O-Ring Specifications:0-8mm: #108 Tip O-Ring, #110 Shaft O-Ring