Learn More
Beckman Coulter O-ring, neoprene, 1 in for PC tube cap (bottle assembly replacement)
Catalog No. NC2592255
Encompass
Neoprene O-ring designed for use with PC tube caps and bottle assemblies. It provides a resilient seal to help prevent leaks and contamination in laboratory bottle and cap applications.
  • Designed for PC tube cap use.
  • Made from neoprene for chemical resistance and sealing performance.
  • Nominal diameter: 1 in (25.4 mm).
  • Intended as a replacement for bottle and cap assemblies.
Catalog No. NC2592255 Supplier Beckman Coulter Supplier No. 870385
Neoprene O-ring designed for use with PC tube caps and bottle assemblies. It provides a resilient seal to help prevent leaks and contamination in laboratory bottle and cap applications.
  • Designed for PC tube cap use.
  • Made from neoprene for chemical resistance and sealing performance.
  • Nominal diameter: 1 in (25.4 mm).
  • Intended as a replacement for bottle and cap assemblies.