Learn More
Thermo Scientific O-Ring Parofluor 2016
Supplier:  Thermo Scientific 29001313
Encompass
O-ring made of perfluoroelastomer intended for use as a seal or gasket in vacuum, chemical, and high temperature applications provides resistance to a wide range of chemicals and maintains sealing properties at extreme temperatures.
Catalog No. NC0904175