Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), White, 150mm Flange
SDP
Catalog No. 50872306
Encompass_Preferred
O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro (FFKM), FEP encapsulated Silicone and FEP encapsulated S.S. spring. The S.S. spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200°CSilicone: -50 to 230°CPerfluoro (FFKM): -7 to 230°CFEP Encapsulated Silicone: -62 to 205°CFEP Encapsulated Stainless: -250 to 205°C
Catalog No. 50-872-306 Supplier Chemglass Life Sciences Supplier No. CG14783
O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro (FFKM), FEP encapsulated Silicone and FEP encapsulated S.S. spring. The S.S. spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200°CSilicone: -50 to 230°CPerfluoro (FFKM): -7 to 230°CFEP Encapsulated Silicone: -62 to 205°CFEP Encapsulated Stainless: -250 to 205°C