Learn More
Chemglass Life Sciences Chemglass O-ring, Perfluoro (FFKM), for 80 mm reaction vessel flange (CG-147-81)
SDP
Catalog No. NC2398020
Encompass_Preferred
Chemglass CG-147-81 is a perfluoro (FFKM) O-ring designed to seal 80 mm flanges on reaction vessels and lids. The perfluoroelastomer construction provides broad chemical resistance and high temperature stability for demanding laboratory and process applications.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • Designed for 80 mm flange sealing applications.
  • Temperature range: -7 to 230 °C.
  • Color: white as supplied.
  • Suitable for aggressive acids, solvents, and high-temperature chemistries.
Catalog No. NC2398020 Supplier Chemglass Life Sciences Supplier No. CG14781
Chemglass CG-147-81 is a perfluoro (FFKM) O-ring designed to seal 80 mm flanges on reaction vessels and lids. The perfluoroelastomer construction provides broad chemical resistance and high temperature stability for demanding laboratory and process applications.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • Designed for 80 mm flange sealing applications.
  • Temperature range: -7 to 230 °C.
  • Color: white as supplied.
  • Suitable for aggressive acids, solvents, and high-temperature chemistries.