Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring #006 - replacement O-ring for reaction vessels and lids (CG-309-U-006)
SDP
Catalog No. 502743595
Encompass_Preferred
Perfluoro (FFKM) O-ring in size #006 designed for demanding chemical sealing applications. This FFKM elastomer provides broad chemical compatibility and thermal stability, making it suitable as a replacement sealing element for reaction vessels, lids, and compatible laboratory fittings. Rated for a wide service temperature range and supplied in a 75 durometer formulation.
  • Perfluoro (FFKM) elastomer for superior chemical resistance.
  • White color and 75 durometer for consistent sealing performance.
  • Service temperature range -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols, ethers, and many organic solvents.
  • Available in standardized size #006 for compatible fittings and components.
Catalog No. 50-274-3595 Supplier Chemglass Life Sciences Supplier No. CG309U006
Perfluoro (FFKM) O-ring in size #006 designed for demanding chemical sealing applications. This FFKM elastomer provides broad chemical compatibility and thermal stability, making it suitable as a replacement sealing element for reaction vessels, lids, and compatible laboratory fittings. Rated for a wide service temperature range and supplied in a 75 durometer formulation.
  • Perfluoro (FFKM) elastomer for superior chemical resistance.
  • White color and 75 durometer for consistent sealing performance.
  • Service temperature range -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols, ethers, and many organic solvents.
  • Available in standardized size #006 for compatible fittings and components.