Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-Ring #008, White - sealing component for fittings and assemblies
SDP
Catalog No. 502743594
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #008, white, designed for demanding chemical and high-temperature sealing applications.
  • High chemical resistance suitable for aggressive solvents and acids.
  • Effective service temperature: -7 to 230° C.
  • 75 durometer (standard hardness).
  • Perfluoroelastomer material for enhanced thermal stability.
  • Supplied individually (each) for replacement or maintenance use.
Catalog No. 50-274-3594 Supplier Chemglass Life Sciences Supplier No. CG309U008
Perfluoro (FFKM) O-ring, size #008, white, designed for demanding chemical and high-temperature sealing applications.
  • High chemical resistance suitable for aggressive solvents and acids.
  • Effective service temperature: -7 to 230° C.
  • 75 durometer (standard hardness).
  • Perfluoroelastomer material for enhanced thermal stability.
  • Supplied individually (each) for replacement or maintenance use.