Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-Ring, Size #009, White, 75 Durometer
SDP
Catalog No. 502743593
Encompass_Preferred
Perfluoro (FFKM) O-ring size #009 is a white, 75 durometer perfluoroelastomer seal designed for demanding chemical environments. It offers broad chemical compatibility and a wide temperature range, making it suitable for laboratory sealing and replacement applications.
  • Material: perfluoroelastomer (FFKM).
  • Color: white.
  • Durometer: 75 durometer.
  • Temperature range: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes, ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Typical use: replacement sealing O-ring for laboratory glassware fittings and assemblies.
Catalog No. 50-274-3593 Supplier Chemglass Life Sciences Supplier No. CG309U009
Perfluoro (FFKM) O-ring size #009 is a white, 75 durometer perfluoroelastomer seal designed for demanding chemical environments. It offers broad chemical compatibility and a wide temperature range, making it suitable for laboratory sealing and replacement applications.
  • Material: perfluoroelastomer (FFKM).
  • Color: white.
  • Durometer: 75 durometer.
  • Temperature range: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes, ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Typical use: replacement sealing O-ring for laboratory glassware fittings and assemblies.