Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), Size #011, White - Replacement seal for Chemglass fittings and glassware
SDP
Catalog No. 502743592
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #011, provides high thermal stability and broad chemical resistance for demanding laboratory sealing applications. Stocked in 75 durometer and suitable for elevated temperatures and aggressive solvents.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color for easy identification.
  • 75 durometer for durable sealing performance.
  • Service temperature -7 to 230 °C.
  • Recommended for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold individually (each) for replacement and maintenance.
Catalog No. 50-274-3592 Supplier Chemglass Life Sciences Supplier No. CG309U011
Perfluoro (FFKM) O-ring, size #011, provides high thermal stability and broad chemical resistance for demanding laboratory sealing applications. Stocked in 75 durometer and suitable for elevated temperatures and aggressive solvents.
  • Perfluoro (FFKM) material for superior chemical resistance.
  • White color for easy identification.
  • 75 durometer for durable sealing performance.
  • Service temperature -7 to 230 °C.
  • Recommended for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Sold individually (each) for replacement and maintenance.