Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring #012, White - Replacement for Laboratory Fittings
SDP
Catalog No. 502743591
Encompass_Preferred
Chemglass Perfluoro (FFKM) O-ring, size #012, is a thermally stable, chemically resistant seal intended for replacement and maintenance of laboratory fittings and glassware connections.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • Stocked white, 75 durometer.
  • Effective service temperature -7 to 230 °C.
  • Sold individually as a single o-ring.
  • Suitable for acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Intended for use as a replacement seal in laboratory fittings and glassware connections.
Catalog No. 50-274-3591 Supplier Chemglass Life Sciences Supplier No. CG309U012
Chemglass Perfluoro (FFKM) O-ring, size #012, is a thermally stable, chemically resistant seal intended for replacement and maintenance of laboratory fittings and glassware connections.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • Stocked white, 75 durometer.
  • Effective service temperature -7 to 230 °C.
  • Sold individually as a single o-ring.
  • Suitable for acids, inorganic bases, alcohols/ethers, aromatic and aliphatic hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Intended for use as a replacement seal in laboratory fittings and glassware connections.