Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, #013, White (CG-309-U-013)
SDP
Catalog No. 502743590
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #013, white. Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance and thermal stability for demanding laboratory sealing applications.
  • Material: Perfluoro (FFKM).
  • Size/number: #013.
  • Color: White.
  • Temperature range: -7 to 230° C.
  • Durometer: 75 durometer.
  • Packaging: Each.
Catalog No. 50-274-3590 Supplier Chemglass Life Sciences Supplier No. CG309U013
Perfluoro (FFKM) O-ring, size #013, white. Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance and thermal stability for demanding laboratory sealing applications.
  • Material: Perfluoro (FFKM).
  • Size/number: #013.
  • Color: White.
  • Temperature range: -7 to 230° C.
  • Durometer: 75 durometer.
  • Packaging: Each.