Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, #014, White
SDP
Catalog No. 502743589
Encompass_Preferred
Chemglass CG-309-U-014 is a perfluoroelastomer (FFKM) o-ring in size #014, supplied in white at 75 durometer. It is designed for high chemical resistance and thermal stability for demanding laboratory and process applications.
  • Perfluoro (FFKM) elastomer material.
  • Stocked in white, 75 durometer.
  • Effective service temperature range of -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols and ethers.
  • Compatible with aromatic hydrocarbons, esters, aldehydes and ketones.
  • Resistant to aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Call for current availability.
Catalog No. 50-274-3589 Supplier Chemglass Life Sciences Supplier No. CG309U014
Chemglass CG-309-U-014 is a perfluoroelastomer (FFKM) o-ring in size #014, supplied in white at 75 durometer. It is designed for high chemical resistance and thermal stability for demanding laboratory and process applications.
  • Perfluoro (FFKM) elastomer material.
  • Stocked in white, 75 durometer.
  • Effective service temperature range of -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols and ethers.
  • Compatible with aromatic hydrocarbons, esters, aldehydes and ketones.
  • Resistant to aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Call for current availability.