Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #015, White (CG-309-U-015)
SDP
Catalog No. 502743588
Encompass_Preferred
Chemglass O-Ring CG-309-U-015 is a perfluoroelastomer (FFKM) sealing ring in size #015 designed for demanding chemical and thermal environments. It provides broad chemical compatibility and high thermal stability for laboratory and process sealing applications.
  • Material: perfluoroelastomer (FFKM).
  • Color: white.
  • Size: #015.
  • Durometer: 75 (stocked).
  • Effective service temperature: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Benefit: superior chemical resistance and thermal stability for reliable sealing.
Catalog No. 50-274-3588 Supplier Chemglass Life Sciences Supplier No. CG309U015
Chemglass O-Ring CG-309-U-015 is a perfluoroelastomer (FFKM) sealing ring in size #015 designed for demanding chemical and thermal environments. It provides broad chemical compatibility and high thermal stability for laboratory and process sealing applications.
  • Material: perfluoroelastomer (FFKM).
  • Color: white.
  • Size: #015.
  • Durometer: 75 (stocked).
  • Effective service temperature: -7 to 230 °C.
  • Chemical compatibility: acids, inorganic bases, alcohols, ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Benefit: superior chemical resistance and thermal stability for reliable sealing.