Learn More
Chemglass Life Sciences Perfluoro (FFKM) O-Ring #016, White - Chemical-Resistant Seal for Glassware Joints and Pressure Vessels
SDP
Catalog No. 502743587
Encompass_Preferred
Perfluoro (FFKM) O-ring size #016 is a high-performance perfluoroelastomer sealing ring that provides superior chemical resistance and a wide service temperature range. Stocked in white at 75 durometer, it is intended for demanding applications requiring durable, chemically resistant seals.
  • High chemical resistance to acids, bases, alcohols, ethers, and many solvents.
  • Effective service temperature: -7 to 230° C.
  • 75 durometer hardness for long-lasting sealing performance.
  • Compatible with aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable for use in glassware joints, vacuum traps, and pressure vessels where aggressive chemicals are present.
Catalog No. 50-274-3587 Supplier Chemglass Life Sciences Supplier No. CG309U016
Perfluoro (FFKM) O-ring size #016 is a high-performance perfluoroelastomer sealing ring that provides superior chemical resistance and a wide service temperature range. Stocked in white at 75 durometer, it is intended for demanding applications requiring durable, chemically resistant seals.
  • High chemical resistance to acids, bases, alcohols, ethers, and many solvents.
  • Effective service temperature: -7 to 230° C.
  • 75 durometer hardness for long-lasting sealing performance.
  • Compatible with aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Suitable for use in glassware joints, vacuum traps, and pressure vessels where aggressive chemicals are present.