Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #018, White
SDP
Catalog No. 502743586
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #018, provides high chemical resistance and thermal stability for sealing applications in aggressive chemical environments. It is formulated in a white 75-durometer perfluoroelastomer and is suitable for use in laboratory glassware joints, valves, pumps, and fittings where broad solvent compatibility is required.
  • Material: perfluoroelastomer (FFKM).
  • Color: white.
  • Size: #018.
  • Durometer: 75 durometer.
  • Temperature range: -7 to 230° C.
  • Compatible with: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Typical uses: sealing joints, valves, pumps, and fittings in chemically aggressive environments.
Catalog No. 50-274-3586 Supplier Chemglass Life Sciences Supplier No. CG309U018
Perfluoro (FFKM) O-ring, size #018, provides high chemical resistance and thermal stability for sealing applications in aggressive chemical environments. It is formulated in a white 75-durometer perfluoroelastomer and is suitable for use in laboratory glassware joints, valves, pumps, and fittings where broad solvent compatibility is required.
  • Material: perfluoroelastomer (FFKM).
  • Color: white.
  • Size: #018.
  • Durometer: 75 durometer.
  • Temperature range: -7 to 230° C.
  • Compatible with: acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, chlorocarbon solvents.
  • Typical uses: sealing joints, valves, pumps, and fittings in chemically aggressive environments.