Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), Size #019, White - Replacement seal for glassware and fittings
SDP
Catalog No. 502743585
Encompass_Preferred
Perfluoro (FFKM) o-ring designed for demanding chemical environments. The perfluoroelastomer construction provides superior chemical resistance and thermal stability, with a standard 75 durometer hardness and an effective service temperature range of -7 to 230° C. Suitable for sealing applications with a wide range of laboratory reagents and solvents.
  • Provides superior chemical resistance to acids, bases, and organic solvents.
  • High thermal stability for use between -7 and 230° C.
  • White perfluoroelastomer material for long-term chemical compatibility.
  • Standard 75 durometer hardness for reliable sealing performance.
  • Suitable for use in laboratory glassware and fittings exposed to aggressive chemicals.
Catalog No. 50-274-3585 Supplier Chemglass Life Sciences Supplier No. CG309U019
Perfluoro (FFKM) o-ring designed for demanding chemical environments. The perfluoroelastomer construction provides superior chemical resistance and thermal stability, with a standard 75 durometer hardness and an effective service temperature range of -7 to 230° C. Suitable for sealing applications with a wide range of laboratory reagents and solvents.
  • Provides superior chemical resistance to acids, bases, and organic solvents.
  • High thermal stability for use between -7 and 230° C.
  • White perfluoroelastomer material for long-term chemical compatibility.
  • Standard 75 durometer hardness for reliable sealing performance.
  • Suitable for use in laboratory glassware and fittings exposed to aggressive chemicals.