Learn More
Chemglass Life Sciences O-Ring, Perfluoro (FFKM), #020, White - Sealing O-ring
SDP
Catalog No. 502743584
Encompass_Preferred
Chemglass perfluoro (FFKM) O-ring, size #020, white, is designed for chemical-resistant sealing applications. The white 75 durometer FFKM compound provides broad chemical compatibility and high temperature stability for demanding lab and process environments.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • White color, 75 durometer.
  • Size: #020 sealing O-ring.
  • Effective service temperature: -7 to 230 °C.
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
Catalog No. 50-274-3584 Supplier Chemglass Life Sciences Supplier No. CG309U020
Chemglass perfluoro (FFKM) O-ring, size #020, white, is designed for chemical-resistant sealing applications. The white 75 durometer FFKM compound provides broad chemical compatibility and high temperature stability for demanding lab and process environments.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • White color, 75 durometer.
  • Size: #020 sealing O-ring.
  • Effective service temperature: -7 to 230 °C.
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.