Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #021, White
SDP
Catalog No. 502743583
Encompass_Preferred
Perfluoro (FFKM) O-ring #021, white, provides high chemical and thermal resistance for laboratory sealing applications. Stocked in 75 durometer and sold individually.
  • Material: Perfluoro (FFKM).
  • Color: White.
  • Size: #021.
  • Durometer: 75 (stocked).
  • Temperature range: -7 to 230 °C.
  • Packaging: Sold individually (each).
  • Suitable for acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
Catalog No. 50-274-3583 Supplier Chemglass Life Sciences Supplier No. CG309U021
Perfluoro (FFKM) O-ring #021, white, provides high chemical and thermal resistance for laboratory sealing applications. Stocked in 75 durometer and sold individually.
  • Material: Perfluoro (FFKM).
  • Color: White.
  • Size: #021.
  • Durometer: 75 (stocked).
  • Temperature range: -7 to 230 °C.
  • Packaging: Sold individually (each).
  • Suitable for acids, inorganic bases, alcohols and ethers, aromatic hydrocarbons, esters, aldehydes and ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.