Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring, #022, White - Replacement seal for laboratory fittings, glassware joints, and valves
SDP
Catalog No. 502743582
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #022, supplied white at 75 durometer. Perfluoro elastomers provide high thermal stability and broad chemical resistance, making them suitable for demanding laboratory sealing applications.
  • Perfluoro (FFKM) elastomer material.
  • White color, 75 durometer.
  • Service temperature range: -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Packaged and sold individually (each).
Catalog No. 50-274-3582 Supplier Chemglass Life Sciences Supplier No. CG309U022
Perfluoro (FFKM) O-ring, size #022, supplied white at 75 durometer. Perfluoro elastomers provide high thermal stability and broad chemical resistance, making them suitable for demanding laboratory sealing applications.
  • Perfluoro (FFKM) elastomer material.
  • White color, 75 durometer.
  • Service temperature range: -7 to 230° C.
  • Suitable for acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
  • Packaged and sold individually (each).