Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), Size #023, White, For Laboratory O-Ring Joints And Fittings
SDP
Catalog No. 502743581
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #023, white, designed for demanding laboratory sealing applications where aggressive chemicals and elevated temperatures are present. The perfluoroelastomer construction provides superior chemical resistance and maintains sealing performance across a wide temperature range.
  • Perfluoro (FFKM) material for broad chemical compatibility.
  • High thermal stability; effective service temperature -7 to 230 °C.
  • Typical hardness 75 durometer for durable sealing performance.
  • Suitable for acids, bases, alcohols, ethers, hydrocarbons, esters, ketones, amines, and chlorocarbon solvents.
  • Intended for use with laboratory O-ring joints and fittings.
Catalog No. 50-274-3581 Supplier Chemglass Life Sciences Supplier No. CG309U023
Perfluoro (FFKM) O-ring, size #023, white, designed for demanding laboratory sealing applications where aggressive chemicals and elevated temperatures are present. The perfluoroelastomer construction provides superior chemical resistance and maintains sealing performance across a wide temperature range.
  • Perfluoro (FFKM) material for broad chemical compatibility.
  • High thermal stability; effective service temperature -7 to 230 °C.
  • Typical hardness 75 durometer for durable sealing performance.
  • Suitable for acids, bases, alcohols, ethers, hydrocarbons, esters, ketones, amines, and chlorocarbon solvents.
  • Intended for use with laboratory O-ring joints and fittings.