Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-ring, Size #024, White (CG-309-U-024)
SDP
Catalog No. 502743580
Encompass_Preferred
The Chemglass Perfluoro (FFKM) O-ring, size #024 (CG-309-U-024), is a white perfluoroelastomer seal engineered for high thermal stability and broad chemical resistance. It is suitable for reaction vessels, lids, and laboratory equipment exposed to aggressive solvents and corrosive media. The effective service temperature range is -7 to 230° C.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • White color for easy visual identification.
  • Service temperature range: -7 to 230° C.
  • Compatible with acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Supplied individually for replacement and maintenance.
Catalog No. 50-274-3580 Supplier Chemglass Life Sciences Supplier No. CG309U024
The Chemglass Perfluoro (FFKM) O-ring, size #024 (CG-309-U-024), is a white perfluoroelastomer seal engineered for high thermal stability and broad chemical resistance. It is suitable for reaction vessels, lids, and laboratory equipment exposed to aggressive solvents and corrosive media. The effective service temperature range is -7 to 230° C.
  • Perfluoroelastomer (FFKM) material for superior chemical resistance.
  • White color for easy visual identification.
  • Service temperature range: -7 to 230° C.
  • Compatible with acids, inorganic bases, alcohols/ethers, hydrocarbons, esters, aldehydes/ketones, amines, and chlorocarbon solvents.
  • Supplied individually for replacement and maintenance.