Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), #026, White
SDP
Catalog No. 502743579
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #026, white, with an effective service temperature range of -7 to 230° C.
  • Perfluoro (FFKM) material.
  • White color.
  • Effective service temperature: -7 to 230° C.
  • Size designation: #026.
Catalog No. 50-274-3579 Supplier Chemglass Life Sciences Supplier No. CG309U026
Perfluoro (FFKM) O-ring, size #026, white, with an effective service temperature range of -7 to 230° C.
  • Perfluoro (FFKM) material.
  • White color.
  • Effective service temperature: -7 to 230° C.
  • Size designation: #026.