Learn More
Chemglass Life Sciences Chemglass O-Ring, Perfluoro (FFKM), White, Size #027
SDP
Catalog No. 502743578
Encompass_Preferred
Perfluoro (FFKM) O-ring, size #027, for sealing applications requiring broad chemical compatibility and high thermal stability. Manufactured from perfluoroelastomer and stocked in white, 75 durometer.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Suitable for acids, bases, alcohols, ethers, hydrocarbons, esters, aldehydes, ketones, and amines.
  • Stocked in white with approximately 75 durometer hardness.
  • Used for sealing laboratory glassware and chemical systems requiring high thermal stability.
  • Sold individually (each).
Catalog No. 50-274-3578 Supplier Chemglass Life Sciences Supplier No. CG309U027
Perfluoro (FFKM) O-ring, size #027, for sealing applications requiring broad chemical compatibility and high thermal stability. Manufactured from perfluoroelastomer and stocked in white, 75 durometer.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Suitable for acids, bases, alcohols, ethers, hydrocarbons, esters, aldehydes, ketones, and amines.
  • Stocked in white with approximately 75 durometer hardness.
  • Used for sealing laboratory glassware and chemical systems requiring high thermal stability.
  • Sold individually (each).