Learn More
Chemglass Life Sciences Chemglass Perfluoro (FFKM) O-Ring #028, white, 75 durometer
SDP
Catalog No. 502743577
Encompass_Preferred
The Chemglass perfluoro (FFKM) O-ring #028 is a white perfluoroelastomer sealing component designed for laboratory applications that require broad chemical compatibility and high temperature resistance.
  • Made from perfluoroelastomer (FFKM).
  • White color for identification.
  • Size: dash number 028.
  • 75 durometer hardness.
  • Effective service temperature: -7 to 230 °C.
  • Sold individually.
Catalog No. 50-274-3577 Supplier Chemglass Life Sciences Supplier No. CG309U028
The Chemglass perfluoro (FFKM) O-ring #028 is a white perfluoroelastomer sealing component designed for laboratory applications that require broad chemical compatibility and high temperature resistance.
  • Made from perfluoroelastomer (FFKM).
  • White color for identification.
  • Size: dash number 028.
  • 75 durometer hardness.
  • Effective service temperature: -7 to 230 °C.
  • Sold individually.