Learn More
Chemglass Life Sciences Chemglass perfluoro (FFKM) O-ring, size #030, white
SDP
Catalog No. 502743576
Encompass_Preferred
Perfluoro (FFKM) O-ring designed for high thermal stability and broad chemical resistance in sealing applications. This white O-ring in size #030 is suitable as a replacement seal in laboratory glassware assemblies and fluid-handling fittings requiring perfluoroelastomer performance.
  • Superior chemical resistance across a wide range of solvents and reagents.
  • High thermal stability with effective service temperature of -7 to 230° C.
  • Cost-effective alternative to other high-performance perfluoroelastomers.
  • White perfluoroelastomer construction for durability in aggressive chemical environments.
  • Available in size #030 for compatible fittings and seals.
Catalog No. 50-274-3576 Supplier Chemglass Life Sciences Supplier No. CG309U030
Perfluoro (FFKM) O-ring designed for high thermal stability and broad chemical resistance in sealing applications. This white O-ring in size #030 is suitable as a replacement seal in laboratory glassware assemblies and fluid-handling fittings requiring perfluoroelastomer performance.
  • Superior chemical resistance across a wide range of solvents and reagents.
  • High thermal stability with effective service temperature of -7 to 230° C.
  • Cost-effective alternative to other high-performance perfluoroelastomers.
  • White perfluoroelastomer construction for durability in aggressive chemical environments.
  • Available in size #030 for compatible fittings and seals.